3,5-Difluoroisonicotinic acid ≥95%

Supplier: Matrix Scientific

047381-5G
101879-746EA 121.54 USD
101879-746
3,5-Difluoroisonicotinic acid ≥95%
3,5-Difluoroisonicotinic acid

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.0900 g/mol
MDL Number: MFCD04972829
CAS Number: 903522-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR