Ethyl-4-methylthiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylthiazole-5-carboxylic acid ethyl esterEthyl 4-methylthiazole-5-carboxylate

047379-1G
101879-734EA 23.08 USD
101879-734
Ethyl-4-methylthiazole-5-carboxylate ≥95%
Ethyl-4-methylthiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MW: 171.214 g/mol
MDL Number: MFCD00626873
CAS Number: 20582-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR