2,4-Dichloro-N-methoxy-N-methylbenzenecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
2,4-Dichloro-N-methoxy-N-methylbenzamide
047268-500MG
047268-5G
101879-208EA
806.15
USD
101879-208
101879-204
2,4-Dichloro-N-methoxy-N-methylbenzenecarboxamide ≥95%
2,4-Dichloro-N-methoxy-N-methylbenzenecarboxamide
Hazard: Irritant
Formula:
C₉H₉Cl₂NO₂ MW: 234.08 g/mol |
MDL Number:
MFCD08689759 CAS Number: 646528-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...