2-(Benzylamino)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

047256-1G
101879-144EA 284.62 USD
101879-144
2-(Benzylamino)-4-thiazolecarboxylic acid ≥95%
2-(Benzylamino)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂S
MDL Number: MFCD06655105
CAS Number: 165682-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR