N-Ethyl-2-(methylamino)acetamide hydrochloride ≥95%

Supplier: Matrix Scientific

047234-1G
101879-036EA 349.6 USD
101879-036
N-Ethyl-2-(methylamino)acetamide hydrochloride ≥95%
N-Ethyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O
MDL Number: MFCD08060121
CAS Number: 909191-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR