3-(tert-Butyl)-1,2,4-thiadiazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-(tert-butyl)-1,2,4-thiadiazole

047227-500MG
101879-012EA 558.9 USD
101879-012
3-(tert-Butyl)-1,2,4-thiadiazol-5-amine ≥95%
3-(tert-Butyl)-1,2,4-thiadiazol-5-amine

Hazard: Irritant

Formula: C₆H₁₁N₃S
MW: 157.24 g/mol
MDL Number: MFCD09865006
CAS Number: 13383-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR