2,4-Dichloro-6-methylpyridine ≥95%

Supplier: Matrix Scientific

047212-1G
101878-942EA 38.46 USD
101878-942
2,4-Dichloro-6-methylpyridine ≥95%
2,4-Dichloro-6-methylpyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
MW: 162.01 g/mol
MDL Number: MFCD08669834
CAS Number: 42779-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR