tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Ethoxy-3-oxopropanoyl)piperidine, N-BOC protected

047107-1G
101878-492EA 80.5 USD
101878-492
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-(3-ethoxy-3-oxopropanoyl)tetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₅NO₅
MW: 299.37 g/mol
MDL Number: MFCD04115569
CAS Number: 479630-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR