tert-Butyl-3-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-1H-pyrrolo[2,3-b]pyridine, N1-BOC protectedtert-Butyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

047090-500MG 047090-5G 047090-1G
101878-412EA 492.31 USD
101878-412 101878-410 101878-408
tert-Butyl-3-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
tert-Butyl-3-bromo-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃BrN₂O₂
MW: 297.1500 g/mol
MDL Number: MFCD08689737
CAS Number: 226085-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR