4-Amino-3-fluorobenzylalcohol ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-Amino-3-fluorophenyl)methanol
047024-5G
047024-1G
047024-500MG
101878-088EA
1030.77
USD
101878-088
101878-086
101878-084
4-Amino-3-fluorobenzylalcohol ≥95%
4-Amino-3-fluorobenzylalkohol
Hazard: Irritant
Formula:
C₇H₈FNO MW: 141.1429 g/mol |
MDL Number:
MFCD03094241 CAS Number: 146019-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...