Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,6-dichloro-4-(trifluoromethyl)nicotinate

046977-500MG 046977-1G
101877-882EA 658.46 USD
101877-882 101877-884
Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate ≥95%
Methyl-2,6-dichloro-4-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₈H₄Cl₂F₃NO₂
MW: 274.03 g/mol
MDL Number: MFCD12025929
CAS Number: 1130344-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR