2-Bromo-5-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-5-nitrobenzenecarbaldehyde
046941-1G
046941-25G
046941-5G
101877-726EA
17.25
USD
101877-726
101877-728
102551-350
2-Bromo-5-nitrobenzaldehyde ≥95%
2-Bromo-5-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₇H₄BrNO₃ MW: 230.0200 g/mol |
MDL Number:
MFCD00462865 CAS Number: 84459-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...