N-(2-Hydroxyethyl)benzenesulfonamide ≥95%
Supplier: Matrix Scientific
046937-1G
046937-500MG
046937-5G
101877-704EA
181.54
USD
101877-704
101877-706
101877-708
N-(2-Hydroxyethyl)benzenesulfonamide ≥95%
N-(2-Hydroxyethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₈H₁₁NO₃S |
MDL Number:
MFCD00456926 CAS Number: 59724-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...