3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol ≥95%

Supplier: Matrix Scientific

046820-500MG
101877-140EA 1109.23 USD
101877-140
3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol ≥95%
3,4-Dihydro-2H-1,4-benzoxazin-6-ylmethanol

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD11848182
CAS Number: 915160-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR