Methyl-1-cyanocyclohexanecarboxylate ≥95%

Supplier: Matrix Scientific

046788-1G 046788-5G 046788-500MG
101877-010EA 278.46 USD
101877-010 101877-012 101877-014
Methyl-1-cyanocyclohexanecarboxylate ≥95%
Methyl-1-cyanocyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD09030934
CAS Number: 58920-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR