4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
046745-1G
046745-500MG
101876-836EA
350.77
USD
101876-836
101876-834
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde ≥95%
4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₈F₃NOS |
MDL Number:
MFCD04115404 CAS Number: 438577-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...