2-Hydrazino-1,3-benzothiazole-6-carbohydrazide ≥95%
Supplier: Matrix Scientific
046728-5G
046728-500MG
046728-1G
101876-758EA
1600
USD
101876-758
101876-756
101876-754
2-Hydrazino-1,3-benzothiazole-6-carbohydrazide ≥95%
2-Hydrazino-1,3-benzothiazole-6-carbohydrazide
Hazard: Irritant
Formula:
C₈H₉N₅OS |
MDL Number:
MFCD08444017 CAS Number: 924869-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...