tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(3,4-Dichlorobenzyl)-3-formylpiperidine, N-BOC protected

046675-500MG
101876-504EA 1109.23 USD
101876-504
tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-3-(3,4-dichlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₈H₂₃Cl₂NO₃
MW: 372.29 g/mol
MDL Number: MFCD09607941
CAS Number: 952183-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR