2-Methoxy-4,6-dimethylnicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4,6-Dimethyl-2-methoxynicotinonitrile

046661-5G 046661-1G
101876-458EA 149.5 USD
101876-458 101876-456
2-Methoxy-4,6-dimethylnicotinonitrile ≥95%
2-Methoxy-4,6-dimethylnicotinonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
MDL Number: MFCD00124716
CAS Number: 65515-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR