Fmoc-3-amino-3-phenylpropionic acid

Supplier: Matrix Scientific

Synonyms: 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid

042190-1G 042190-5G
101854-826EA 210.77 USD
101854-826 101854-828
Fmoc-3-amino-3-phenylpropionic acid
Fmoc-3-amino-3-phenylpropionic acid

Hazard: Irritant

Formula: C₂₄H₂₁NO₄
MW: 387.44 g/mol
MDL Number: MFCD01863192
CAS Number: 180181-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR