Boc-4-aminocinnamic acid predominantly trans
Supplier: Matrix Scientific
042175-5G
042175-1G
101854-766EA
461.54
USD
101854-766
101854-768
Boc-4-aminocinnamic acid predominantly trans
Boc-4-aminocinnamic acid
Hazard: Irritant
Formula:
C₁₄H₁₇NO₄ |
MDL Number:
MFCD02682305 CAS Number: 197073-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...