Boc-(2-aminomethylphenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(Boc-aminomethyl)phenylacetic Acid

042162-1G 042162-5G
101854-720EA 2207.69 USD
101854-720 101854-718
Boc-(2-aminomethylphenyl)acetic acid
Boc-(2-aminomethylphenyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD06656428
CAS Number: 40851-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR