Boc-L-1,2,3,4-tetrahydronorharman-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (S)-2-(tert-Butoxycarbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid

042136-1G 042136-5G
101854-614EA 113.85 USD
101854-614 101854-616
Boc-L-1,2,3,4-tetrahydronorharman-3-carboxylic acid
Boc-L-1,2,3,4-tetrahydronorharman-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₄
MW: 316.36 g/mol
MDL Number: MFCD00800580
CAS Number: 66863-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR