Boc-α-methyl-DL-tryptophan

Supplier: Matrix Scientific

042132-1G
101854-600EA 920 USD
101854-600
Boc-α-methyl-DL-tryptophan
Boc-alpha-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD02682649
CAS Number: 114779-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR