Boc-7-methyl-DL-tryptophan

Supplier: Matrix Scientific

042131-1G
101854-596EA 2106.15 USD
101854-596
Boc-7-methyl-DL-tryptophan
Boc-7-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD02682378
CAS Number: 1219333-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR