Boc-4-methyl-DL-tryptophan

Supplier: Matrix Scientific

042113-100MG 042113-1G
101854-522EA 412.31 USD
101854-522 101854-524
Boc-4-methyl-DL-tryptophan
Boc-4-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD09750498
CAS Number: 1219232-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR