Fmoc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid

Supplier: Matrix Scientific

042099-1G
101854-466EA 1644.5 USD
101854-466
Fmoc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid
Fmoc-(RS)-2-amino-3-hydroxy-3-methylbutanoic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₅
MDL Number: MFCD02682579
CAS Number: 1246172-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR