Fmoc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Supplier: Matrix Scientific
042093-500MG
042093-100MG
101854-436EA
735.38
USD
101854-436
101854-438
Fmoc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Fmoc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Hazard: Irritant
Formula:
C₂₁H₂₃NO₅ |
MDL Number:
MFCD02682571 CAS Number: 1279034-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...