Boc-D-α-methylserine

Supplier: Matrix Scientific

042085-100MG 042085-1G
101854-400EA 1430.77 USD
101854-400 101854-398
Boc-D-α-methylserine
Boc-D-alpha-methylserine

Hazard: Irritant

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
MDL Number: MFCD02682596
CAS Number: 84311-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR