Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Supplier: Matrix Scientific
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid
, 3-Hydroxy-D-valine, N-BOC protected
042074-100MG
042074-500MG
101854-352EA
275.38
USD
101854-352
101854-354
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₉NO₅ MW: 233.27 g/mol |
MDL Number:
MFCD02682576 CAS Number: 288159-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...