Boc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Supplier: Matrix Scientific
042069-500MG
042069-100MG
101854-330EA
735.38
USD
101854-330
101854-332
Boc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Boc-(2S,3S)-2-amino-3-ethoxybutanoic acid
Hazard: Irritant
Formula:
C₁₁H₂₁NO₅ |
MDL Number:
MFCD02682570 CAS Number: 630422-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...