Fmoc-(R)-5,5-dimethylthiazolidine-4-carboxylicacid

Supplier: Matrix Scientific

042053-500MG 042053-1G
101854-258EA 547.69 USD
101854-258 101854-260
Fmoc-(R)-5,5-dimethylthiazolidine-4-carboxylicacid
Fmoc-(R)-5,5-dimethylthiazolidine-4-carboxylicacid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄S
MDL Number: MFCD02682349
CAS Number: 753030-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR