Fmoc-(2S,5R)-5-phenylpyrrolidine-2-carboxylicacid
Supplier: Matrix Scientific
042049-1G
042049-250MG
101854-242EA
538.46
USD
101854-242
101854-244
Fmoc-(2S,5R)-5-phenylpyrrolidine-2-carboxylicacid
Fmoc-(2S,5R)-5-phenylpyrrolidine-2-carboxylicacid
Hazard: Irritant
Formula:
C₂₆H₂₃NO₄ |
MDL Number:
MFCD00673788 CAS Number: 215190-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...