Fmoc-(2S,3R)-3-phenylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

042042-100MG 042042-250MG
101854-216EA 792.31 USD
101854-216 101854-218
Fmoc-(2S,3R)-3-phenylpyrrolidine-2-carboxylic acid
Fmoc-(2S,3R)-3-phenylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₂₆H₂₃NO₄
MDL Number: MFCD01860724
CAS Number: 281655-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR