Boc-trans-4-hydroxy-D-proline

Supplier: Matrix Scientific

Synonyms: (2R,4S)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid

042038-1G
101854-202EA 166.75 USD
101854-202
Boc-trans-4-hydroxy-D-proline
Boc-trans-4-hydroxy-D-proline

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
MDL Number: MFCD01861341
CAS Number: 147266-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR