Fmoc-O-ethyl-L-tyrosine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid

042009-1G 042009-5G
101854-084EA 323.08 USD
101854-084 101854-082
Fmoc-O-ethyl-L-tyrosine
Fmoc-O-ethyl-L-tyrosine

Hazard: Irritant

Formula: C₂₆H₂₅NO₅
MW: 431.49 g/mol
MDL Number: MFCD02094093
CAS Number: 119894-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR