Fmoc-4-amino-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-D-Phe(4-NH2)-OH
041972-1G
041972-5G
101853-936EA
138.46
USD
101853-936
101853-934
Fmoc-4-amino-D-phenylalanine
Fmoc-4-amino-D-phenylalanine
Hazard: Irritant
Formula:
C₂₄H₂₂N₂O₄ MW: 402.45 g/mol |
MDL Number:
MFCD02682658 CAS Number: 324017-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...