Fmoc-4-(trifluoromethyl)-D-phenylalanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-D-Phe(4-CF3)-OH
041969-5G
041969-1G
101853-924EA
4004.62
USD
101853-924
101853-922
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Fmoc-4-(trifluoromethyl)-D-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₀F₃NO₄ MW: 455.44 g/mol |
MDL Number:
MFCD00797582 CAS Number: 238742-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...