Boc-D-Tyr(Et)-OH

Supplier: Matrix Scientific

Synonyms: N-Boc-O-ethyl-D-tyrosine

041911-1G 041911-5G
101853-700EA 366.15 USD
101853-700 101853-698
Boc-D-Tyr(Et)-OH
Boc-D-Tyr(Et)-OH

CAS: 76757-92-1
MDL Number: MFCD00065602
Molecular Formula: C16H23NO5
Molecular Weight: 309,37
Hazard: Irritant

Formula: C₁₆H₂₃NO₅
MDL Number: MFCD00065602
CAS Number: 76757-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR