(S)-N-Boc-4-bromophenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-p-bromo-Phe-OHBoc-Phe(4-Br)-OH

041882-5G
101853-580EA 78.2 USD
101853-580
(S)-N-Boc-4-bromophenylalanine
(S)-N-Boc-4-bromophenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₄
MW: 344.2 g/mol
MDL Number: MFCD00237571
CAS Number: 62129-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR