Boc-Bpa-OH
Supplier: Matrix Scientific
Synonyms:
Boc-p-Bz-Phe-OH
041879-1G
101853-568EA
189.75
USD
101853-568
Boc-Bpa-OH
Boc-Bpa-OH
CAS: 104504-43-0
MDL Number: MFCD00151886
Molecular Formula: C21H23NO5
Molecular Weight: 369,42
Hazard: Irritant
Formula:
C₂₁H₂₃NO₅ MW: 369.417 g/mol |
MDL Number:
MFCD00151886 CAS Number: 104504-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...