N-Boc-3-methyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3-Me)-OH

041869-1G 041869-5G
101853-528EA 604.62 USD
101853-528 101853-526
N-Boc-3-methyl-L-phenylalanine
N-Boc-3-methyl-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD01862944
CAS Number: 114873-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR