Boc-3-bromo-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-Phe(3-Br)-OH

041854-1G
101853-472EA 54.05 USD
101853-472
Boc-3-bromo-L-phenylalanine
Boc-3-bromo-L-phenylalanine

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₄
MW: 344.21 g/mol
MDL Number: MFCD01317710
CAS Number: 82278-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR