Boc-D-Tic-OH
Supplier: Matrix Scientific
Synonyms:
Boc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
041824-5G
041824-1G
101853-352EA
216.92
USD
101853-352
101853-350
Boc-D-Tic-OH
Boc-D-Tic-OH
CAS: 115962-35-1
MDL Number: MFCD00143818
Molecular Formula: C15H19NO4
Molecular Weight: 277,32
Hazard: Irritant
Formula:
C₁₅H₁₉NO₄ MW: 277.32 g/mol |
MDL Number:
MFCD00143818 CAS Number: 115962-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...