Di-boc-S-(2-aminoethyl)-L-cysteine
Supplier: Matrix Scientific
041780-1G
041780-5G
101853-158EA
409.4
USD
101853-158
101853-160
Di-boc-S-(2-aminoethyl)-L-cysteine
Di-boc-S-(2-aminoethyl)-L-cysteine
Hazard: Irritant
Formula:
C₁₅H₂₈N₂O₆S |
MDL Number:
MFCD12547716 CAS Number: 130622-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...