Boc-Cys(Dpm)-OH

Supplier: Matrix Scientific

Synonyms: Boc-Cys(Dpm)-OHS-Benzhydryl-N-Boc-L-cysteine

041769-5G 041769-1G
101853-116EA 1001.54 USD
101853-116 101853-114
Boc-Cys(Dpm)-OH
Boc-Cys(Dpm)-OH

CAS: 21947-97-7
MDL Number: MFCD00151869
Molecular Formula: C21H25NO4S
Molecular Weight: 387,5
Hazard: Irritant

Formula: C₂₁H₂₅NO₄S
MW: 387.494 g/mol
MDL Number: MFCD00151869
CAS Number: 21947-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR