N-Boc-S-benzyl-D-cysteine
Supplier: Matrix Scientific
Synonyms:
Boc-D-Cys(Bzl)-OH
041763-5G
041763-25G
102551-296EA
707.69
USD
102551-296
101853-092
N-Boc-S-benzyl-D-cysteine
N-Boc-S-benzyl-D-cysteine
CAS: 102830-49-9
MDL Number: MFCD00038253
Molecular Formula: C15H21NO4S
Molecular Weight: 311,4
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄S MW: 311.396 g/mol |
MDL Number:
MFCD00038253 CAS Number: 102830-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...