Boc-Pen(pMeBzl)-OH
Supplier: Matrix Scientific
Synonyms:
Boc-Pen(Mbzl)-OH·DCHA
041760-5G
041760-1G
101853-080EA
1026.15
USD
101853-080
101853-078
Boc-Pen(pMeBzl)-OH
Boc-Pen(pMeBzl)-OH
CAS: 198474-61-2
MDL Number: MFCD00235907
Molecular Formula: C30H50N2O4S
Molecular Weight: 534,81
Hazard: Irritant
Formula:
C₃₀H₅₀N₂O₄S MW: 534.8 g/mol |
CAS Number:
198474-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...