Boc-DL-buthioninesulfoximine

Supplier: Matrix Scientific

041747-1G 041747-250MG
101853-024EA 571.46 USD
101853-024 101853-022
Boc-DL-buthioninesulfoximine
Boc-DL-buthioninesulfoximine

Hazard: Irritant

Formula: C₁₃H₂₆N₂O₅S
MDL Number: MFCD02682524
CAS Number: 128377-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR