Fmoc-3-Abz-OH

Supplier: Matrix Scientific

041729-1G
101852-944EA 16.1 USD
101852-944
Fmoc-3-Abz-OH
Fmoc-3-Abz-OH

Hazard: Irritant

Formula: C₂₂H₁₇NO₄
MDL Number: MFCD00235885
CAS Number: 185116-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR